Learn More
MSC Miller Rescue Descent Device, Iron and Aluminum, 5 in. x 10 ft., 1 ea.
Catalog No. 5000509002
Encompass
The descent device unit is a manual rescue device designed for single-person controlled descents. It allows the user to manage descent speed through friction by wrapping the work line around an aluminum mandrel, with manual tension on the line providing deceleration and stopping. The device can also be locked into a stationary position to support a hands-free work environment.
  • Designed for single-person use
  • Provides user-controlled descent speed and braking
  • Features a helical grooved contour for work line friction
  • Allows for hands-free stationary positioning
  • Meets applicable OSHA, ANSI and CSA standards
  • Requires use with polyester-braided descent rope
Catalog No. 50-005-09002 Supplier MSC Supplier No. 31210438
The descent device unit is a manual rescue device designed for single-person controlled descents. It allows the user to manage descent speed through friction by wrapping the work line around an aluminum mandrel, with manual tension on the line providing deceleration and stopping. The device can also be locked into a stationary position to support a hands-free work environment.
  • Designed for single-person use
  • Provides user-controlled descent speed and braking
  • Features a helical grooved contour for work line friction
  • Allows for hands-free stationary positioning
  • Meets applicable OSHA, ANSI and CSA standards
  • Requires use with polyester-braided descent rope