Learn More
Medchemexpress LLC HY-42110 100mg , Deschloroclozapine CAS:1977-07-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY42110100MG
Encompass_Preferred
HY-42110 100mg Deschloroclozapine CAS: 1977-07-7 Deschloroclozapine, a metabolite of Clozapine, is a highly potent muscarinic DREADDs agonist. Deschloroclozapine binds to DREADD receptor subtypes hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM, respectively. [11C]-Deschloroclozapine is developed as a promising PET tracer for DREADD imaging. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9052