Learn More
Medchemexpress LLC HY-42110 10mg Medchemexpress, Deschloroclozapine CAS:1977-07-7 Purity:>98%
SDP
Catalog No. 502029315
Encompass_Preferred
Medchemexpress, HY-42110 10mg Deschloroclozapine CAS:1977-07-7 Deschloroclozapine is a potent, high affinity, selective, metabolically stable agonist of muscarinic-based DREADDs. Deschloroclozapine inhibites [3H]quinuclidinyl benzilate (QNB) binding to hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM. Deschloroclozapine is reported with utility in both mice and non-human primates for a variety of applications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9315 Supplier Medchemexpress LLC Supplier No. HY4211010MG
Medchemexpress, HY-42110 10mg Deschloroclozapine CAS:1977-07-7 Deschloroclozapine is a potent, high affinity, selective, metabolically stable agonist of muscarinic-based DREADDs. Deschloroclozapine inhibites [3H]quinuclidinyl benzilate (QNB) binding to hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM. Deschloroclozapine is reported with utility in both mice and non-human primates for a variety of applications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.