Learn More
Medchemexpress LLC Deschloroclozapine dihydrochloride | 99.9% | 365.30 | 100 MG
SDP
Catalog No. 5000403505
Encompass_Preferred
Deschloroclozapine dihydrochloride, a metabolite of Clozapine, is a highly potent muscarinic DREADDs agonist. It binds to DREADD receptor subtypes hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM, respectively. [11C]-Deschloroclozapine is developed as a promising PET tracer for DREADD imaging.
  • Highly potent muscarinic DREADDs agonist.
  • Binds to DREADD receptor subtypes hM3Dq and hM4Di.
  • Developed as a promising PET tracer for DREADD imaging.
Catalog No. 50-004-03505 Supplier Medchemexpress LLC Supplier No. HY42110A100MG
Deschloroclozapine dihydrochloride, a metabolite of Clozapine, is a highly potent muscarinic DREADDs agonist. It binds to DREADD receptor subtypes hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM, respectively. [11C]-Deschloroclozapine is developed as a promising PET tracer for DREADD imaging.
  • Highly potent muscarinic DREADDs agonist.
  • Binds to DREADD receptor subtypes hM3Dq and hM4Di.
  • Developed as a promising PET tracer for DREADD imaging.