Learn More
Medchemexpress LLC Deschloroetizolam | 40054-73-7 | 99.8% | 308.41 | 100 MG
SDP
Catalog No. 5000347909
Encompass_Preferred
Deschloroetizolam is a designer Benzodiazepine, a psychoactive substance with physical and cognitive effects. It is intended for research use only, not for sale to patients. This solid, white to off-white compound has a molecular weight of 308.41. It exhibits high solubility in DMSO, reaching 125 mg/mL (405.30 mM).
  • Intended for research use only
  • Psychoactive substance with physical and cognitive effects
  • Appears as a white to off-white solid
  • Highly soluble in DMSO (125 mg/mL)
Catalog No. 50-003-47909 Supplier Medchemexpress LLC Supplier No. HY137994100MG
Deschloroetizolam is a designer Benzodiazepine, a psychoactive substance with physical and cognitive effects. It is intended for research use only, not for sale to patients. This solid, white to off-white compound has a molecular weight of 308.41. It exhibits high solubility in DMSO, reaching 125 mg/mL (405.30 mM).
  • Intended for research use only
  • Psychoactive substance with physical and cognitive effects
  • Appears as a white to off-white solid
  • Highly soluble in DMSO (125 mg/mL)