Learn More
Medchemexpress LLC Deschloroetizolam | 40054-73-7 | MFCD02942552 | 99.8% | 308.41 g/mol | C17H16N4S | 5 MG
SDP
Catalog No. 5000222936
Encompass_Preferred
Deschloroetizolam is a thienotriazolodiazepine provided as a research and analytical reference material for laboratory and forensic applications. The product is documented with CAS 40054-73-7, molecular formula C17H16N4S, and molecular weight 308.41 g/mol. It is offered in solid and solution formats and is accompanied by a datasheet and SDS; confirm lot-specific certificate of analysis for exact purity and specifications.
  • High reported purity suitable for analytical workflows.
  • Available in small solid quantities and pre-made solutions for convenience.
  • Accompanied by a downloadable datasheet and safety data sheet.
  • Documented molecular formula and molecular weight for traceability.
  • Intended for laboratory and forensic research use only.
Catalog No. 50-002-22936 Supplier Medchemexpress LLC Supplier No. HY1379945MG
Deschloroetizolam is a thienotriazolodiazepine provided as a research and analytical reference material for laboratory and forensic applications. The product is documented with CAS 40054-73-7, molecular formula C17H16N4S, and molecular weight 308.41 g/mol. It is offered in solid and solution formats and is accompanied by a datasheet and SDS; confirm lot-specific certificate of analysis for exact purity and specifications.
  • High reported purity suitable for analytical workflows.
  • Available in small solid quantities and pre-made solutions for convenience.
  • Accompanied by a downloadable datasheet and safety data sheet.
  • Documented molecular formula and molecular weight for traceability.
  • Intended for laboratory and forensic research use only.