Learn More
Medchemexpress LLC HY-107339 5mg Medchemexpress, Deserpidine CAS:131-01-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1073395MG
Encompass_Preferred
Medchemexpress, HY-107339 5mg Deserpidine CAS:131-01-1 Deserpidine (Harmonyl) is an alkaloid isolated from the root of Rauwolfia canescens related to Reserpine. Deserpidine is used as an antihypertensive agent and a tranquilizer. Deserpidine is a competitive angiotensin converting enzyme (ACE) inhibitor. Deserpidine also decreases angiotensin II-induced aldosterone secretion by the adrenal cortex[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8385