Learn More
Desert Assembly Opti-Klens™ Wash Fountain
Catalog No. 19269422
Change view
Click to view available options
Includes:
Diverter, Chrome Plated Brass Valve, Adapters, Dust Cover
Two spray nozzles, diverter, Chrome Plated Brass valve, adapters
Product Type:
Opti-Klens I Eyewash
Opti-Klens II Eyewash
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Product Type
19-269422 Two spray nozzles, diverter, Chrome Plated Brass valve, adapters Opti-Klens II Eyewash
19-269421 Diverter, Chrome Plated Brass Valve, Adapters, Dust Cover Opti-Klens I Eyewash
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-269422 Supplier Desert Assembly Supplier No. OPTIKLENS II
Chrome-plated, solid brass unit easily installs on any existing sink or basin faucet as a permanent eye wash fountain, yet does not interfere with normal faucet functioning. Water supply in an emergency is limitless, unlike portable models. In operation, unit frees both hands to hold eyes open to the flow of water. No plumbing is required. Supplied with two of the most common adapters. Other adapters are available from the manufacturer at no additional cost.
Compliance
ANSI compliant; meet the ANSI Z358.1 standard for an eye wash unit. Spray configuration on Opti-Klens II allows both eyes and face to be washed.
Specifications
Certifications/Compliance ANSI Z358.1-2014
For Use With (Application) Eye and Face Wash
Includes Two spray nozzles, diverter, Chrome Plated Brass valve, adapters
Product Type Opti-Klens II Eyewash