Learn More
Desert Assembly Opti-Klens™ Wash Fountain
Catalog No. p-35163
Click to view available options
Includes:
Diverter, Chrome Plated Brass Valve, Adapters, Dust Cover
Two spray nozzles, diverter, Chrome Plated Brass valve, adapters
Product Type:
Opti-Klens I Eyewash
Opti-Klens II Eyewash
Chrome-plated, solid brass unit easily installs on any existing sink or basin faucet as a permanent eye wash fountain, yet does not interfere with normal faucet functioning. Water supply in an emergency is limitless, unlike portable models. In operation, unit frees both hands to hold eyes open to the flow of water. No plumbing is required. Supplied with two of the most common adapters. Other adapters are available from the manufacturer at no additional cost.
Compliance
ANSI compliant; meet the ANSI Z358.1 standard for an eye wash unit. Spray configuration on Opti-Klens II allows both eyes and face to be washed.
Specifications
Certifications/Compliance ANSI Z358.1-2014
For Use With (Application) Eye and Face Wash
Includes Two spray nozzles, diverter, Chrome Plated Brass valve, adapters
Product Type Opti-Klens II Eyewash
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.