Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desethyl chloroquine diphosphate | 247912-76-1 | MFCD28138115 | 99.7% | 487.81 | C16H28ClN3O8P2 | 5 MG
SDP

Catalog No. 5000222633
Encompass_Preferred

Desethyl chloroquine diphosphate is the diphosphate salt of desethyl chloroquine supplied as a research reagent and analytical standard. It is provided as a high-purity solid intended for biochemical, pharmacological, and analytical applications, with documentation including a data sheet and safety data sheet.

  • High purity suitable for analytical work (reported 99.7%).
  • Diphosphate salt form for improved solubility and stability.
  • Molecular weight 487.81 and formula C16H28ClN3O8P2 for accurate calculations.
  • Available in small milligram pack sizes for reference and method development.
  • Supplied with datasheet and SDS for handling and compliance.

Catalog No. 50-002-22633 Supplier Medchemexpress LLC Supplier No. HY135811A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desethyl chloroquine diphosphate is the diphosphate salt of desethyl chloroquine supplied as a research reagent and analytical standard. It is provided as a high-purity solid intended for biochemical, pharmacological, and analytical applications, with documentation including a data sheet and safety data sheet.

  • High purity suitable for analytical work (reported 99.7%).
  • Diphosphate salt form for improved solubility and stability.
  • Molecular weight 487.81 and formula C16H28ClN3O8P2 for accurate calculations.
  • Available in small milligram pack sizes for reference and method development.
  • Supplied with datasheet and SDS for handling and compliance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.