Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desethyl chloroquine diphosphate | 247912-76-1 | MFCD34469969; MFCD28138115 | 99.7% | C16H28ClN3O8P2 | 10MG
SDP

Catalog No. 5000259441
Encompass_Preferred

Desethyl chloroquine diphosphate is the diphosphate salt of desethyl chloroquine, a primary desethyl metabolite of chloroquine. It is provided as a research-grade antiplasmodic compound for biochemical and pharmacological studies, offered at high purity and in small pack sizes suitable for laboratory experiments.

  • High reported purity (99.7%).
  • Available in small pack sizes, including 10 mg.
  • Supplied as the diphosphate salt.
  • Molecular formula C16H28ClN3O8P2; molecular weight 487.81.
  • Suitable for biochemical and pharmacological research applications.

Catalog No. 50-002-59441 Supplier Medchemexpress LLC Supplier No. HY135811A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desethyl chloroquine diphosphate is the diphosphate salt of desethyl chloroquine, a primary desethyl metabolite of chloroquine. It is provided as a research-grade antiplasmodic compound for biochemical and pharmacological studies, offered at high purity and in small pack sizes suitable for laboratory experiments.

  • High reported purity (99.7%).
  • Available in small pack sizes, including 10 mg.
  • Supplied as the diphosphate salt.
  • Molecular formula C16H28ClN3O8P2; molecular weight 487.81.
  • Suitable for biochemical and pharmacological research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.