Learn More
Medchemexpress LLC Desethyl dabigatran etexilate | 212321-78-3 | 599.68 | 25 MG
SDP
Catalog No. 5000447758
Encompass_Preferred
Desethyl Dabigatran Etexilate is an intermediate metabolite of dabigatran etexilate, primarily used for research purposes. This compound is supplied as a white to yellow solid.
  • Intermediate metabolite
  • For research use only
  • White to yellow solid appearance
  • Stable as powder at -20°C for 3 years, or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-47758 Supplier Medchemexpress LLC Supplier No. 25MGHYW004360
Desethyl Dabigatran Etexilate is an intermediate metabolite of dabigatran etexilate, primarily used for research purposes. This compound is supplied as a white to yellow solid.
  • Intermediate metabolite
  • For research use only
  • White to yellow solid appearance
  • Stable as powder at -20°C for 3 years, or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months, or -20°C for 1 month