Learn More
Cayman Chemical Desethylchloroquine, Purity: ≥95%, CAS: 1476-52-4, 10 mg
SDP
Catalog No. 502990062
Encompass_Preferred
An active metabolite of chloroquine; formed when chloroquine undergoes dealkylation, primarily by CYP2C8 and CYP3A4 and to a lesser extent by CYP2D6; inhibits the growth of the P. falciparum strain LA136 in vitro (IC50 = 9.9 ng/ml)
Catalog No. 50-299-0062 Supplier Cayman Chemical Supplier No. 3011310 MG
An active metabolite of chloroquine; formed when chloroquine undergoes dealkylation, primarily by CYP2C8 and CYP3A4 and to a lesser extent by CYP2D6; inhibits the growth of the P. falciparum strain LA136 in vitro (IC50 = 9.9 ng/ml)