Learn More
Cayman Chemical DesethylchloroquIn-d4 1mg
SDP
Catalog No. 502991928
Encompass_Preferred
An internal standard for the quantification of desethylchloroquine by GC- or LC-MS
Catalog No. 50-299-1928 Supplier Cayman Chemical Supplier No. 309071 MG
An internal standard for the quantification of desethylchloroquine by GC- or LC-MS