Learn More
TARGETMOL CHEMICALS INC DESFORMYLFLUSTRABROMINE 25MG
SDP
Catalog No. 502581603
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Desformylflustrabromine hydrochloride (dFBr hydrochloride) is a selective agonist of nicotinic acetylcholine receptor (nAChR) in (alpha)4(beta)2 neurons with pEC50 of 6.48. purity: 98%
Catalog No. 50-258-1603 Supplier TARGETMOL CHEMICALS INC Supplier No. T1100425MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Desformylflustrabromine hydrochloride (dFBr hydrochloride) is a selective agonist of nicotinic acetylcholine receptor (nAChR) in (alpha)4(beta)2 neurons with pEC50 of 6.48. purity: 98%