Learn More
Medchemexpress LLC HY-107675 5mg Medchemexpress, Desformylflustrabromine hydrochloride CAS:951322-11-5 Purity:>98%
SDP
Catalog No. 501968451
Encompass_Preferred
Medchemexpress, HY-107675 5mg Desformylflustrabromine HCl CAS:951322-11-5 Desformylflustrabromine hydrochloride is a selective agonist of α4β2 neuronal nicotinic acetylcholine receptor (nAChR) with a pEC50 of 6.48. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8451 Supplier Medchemexpress LLC Supplier No. HY1076755MG
Medchemexpress, HY-107675 5mg Desformylflustrabromine HCl CAS:951322-11-5 Desformylflustrabromine hydrochloride is a selective agonist of α4β2 neuronal nicotinic acetylcholine receptor (nAChR) with a pEC50 of 6.48. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.