Learn More
Abcam Desformylflustrabromine hydrochloride, alpha4beta2 modulator, 50MG
Catalog No. 502706173
Encompass_Preferred
MW 357.7 Da, Purity >98%. Selective positive allosteric nAChR α4β2 modulator (Ki = 3400 nM). Increases whole cell current when co-applied with effective concentrations of ACh. Anti-addictive effects in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6173 Supplier Abcam Supplier No. AB14281550MG
Add to Cart
Add to Cart
MW 357.7 Da, Purity >98%. Selective positive allosteric nAChR α4β2 modulator (Ki = 3400 nM). Increases whole cell current when co-applied with effective concentrations of ACh. Anti-addictive effects in vivo..
The product is subject to the following: Abcam Restricted Use Statement