Learn More
Cayman Chemical Desglymidodrine, Purity: ≥98%, CAS: 3600-87-1, 10 mg
SDP
Catalog No. 502913206
Encompass_Preferred
An active metabolite of midodrine; an agonist of α1-ARs; binds to α1-ARs (Kis = 9.77, 9.55, and 8.51 µM for α1A, α1B, and α1D-ARs, respectively); increases IUP and MAP in anesthetized dogs by 5 and 20 mm Hg, respectively, in anesthetized dogs at 205 and 250 nmol/kg, respectively
Catalog No. 50-291-3206 Supplier Cayman Chemical Supplier No. 3560810 MG
An active metabolite of midodrine; an agonist of α1-ARs; binds to α1-ARs (Kis = 9.77, 9.55, and 8.51 µM for α1A, α1B, and α1D-ARs, respectively); increases IUP and MAP in anesthetized dogs by 5 and 20 mm Hg, respectively, in anesthetized dogs at 205 and 250 nmol/kg, respectively