Learn More
Research Products International Corp Desi-Can Desiccant, 2 x 4 x 1/2 Inches
SDP
Catalog No. 50998060
Encompass
Excellent for use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture, these canisters contain silica gel to effectively absorb moisture through perforations in the aluminum case.
•    One canister of Reusable Desi-Can™ maintains a safe level of humidity in 0.09 cubic meter (3 cubic ft.)
      of space.
•    Can be regenerated in a 150ºC (300ºF) oven in about 3 hours for unlimited life.
•    Monitor window shows blue when recharged and pink when saturated.
Catalog No. 50-998-060 Supplier Research Products International Corp Supplier No. 420105
Excellent for use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture, these canisters contain silica gel to effectively absorb moisture through perforations in the aluminum case.
•    One canister of Reusable Desi-Can™ maintains a safe level of humidity in 0.09 cubic meter (3 cubic ft.)
      of space.
•    Can be regenerated in a 150ºC (300ºF) oven in about 3 hours for unlimited life.
•    Monitor window shows blue when recharged and pink when saturated.