Learn More
Waters Corp Service Kit, DESI Source, MS System, Annual Replacement
Catalog No. 502390477 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This service kit is designed for maintaining a DESI Source. It includes replacement fused silica, an o-ring, and a pack of Taper Tip Emitters.
  • Designed as a rebuild kit
  • Compatible with MS systems
  • Recommended for annual replacement
  • Includes replacement fused silica
  • Includes an o-ring
  • Includes a pack of Taper Tip Emitters
Catalog No. 50-239-0477 Supplier Waters Corp Supplier No. 700011424
This service kit is designed for maintaining a DESI Source. It includes replacement fused silica, an o-ring, and a pack of Taper Tip Emitters.
  • Designed as a rebuild kit
  • Compatible with MS systems
  • Recommended for annual replacement
  • Includes replacement fused silica
  • Includes an o-ring
  • Includes a pack of Taper Tip Emitters