Learn More
WA HAMMOND DRIERITE COMPANY DRIERITE, DESICCANT BAGS 2 OZ. KRAFT PAPER 400BAGS/CASE, STOCK# 60012
Catalog No. NC3445036
Encompass
These non-woven bags containing Drierite are used to completely dry the air within a package or container during shipment or storage. The disposable packets are dust tight and will protect enclosures for many months when used within an adequate moisture barrier. These desiccant bags are approved by the FDA and Drierite is Generally Recognized As Safe (GRAS).
Catalog No. NC3445036 Supplier WA HAMMOND DRIERITE COMPANY Supplier No. 60012
These non-woven bags containing Drierite are used to completely dry the air within a package or container during shipment or storage. The disposable packets are dust tight and will protect enclosures for many months when used within an adequate moisture barrier. These desiccant bags are approved by the FDA and Drierite is Generally Recognized As Safe (GRAS).