Learn More
Hach Company Desiccant Cartridge, 6/pk
Supplier:  Hach Company 5188906
Encompass
These silica gel desiccant cartridges are used with the dry storage chamber for the Portable LDO Probe.
Catalog No. 50-199-7136