Learn More
Electron Microscopy Sciences Desiccant Cartridge, Small
SDP
Catalog No. 501899508
Encompass_Preferred
These desiccant cartridges can be used in desiccator cabinets or non-vacuum desiccators, analytical balance cases, and precision laboratory instruments. The clear polystyrene dish allows for instant visual inspection of the silica gel color which changes from blue to pale pink as it absorbs moisture.
A highly porous membrane covers the dish of the desiccant providing good moisture absorption. The desiccant can be regenerated by heating cartridges in an oven at 50 degree C (122 degree F) overnight or in a 150 degree C (302 degree F) oven for one hour. The silica gel is inert and non-toxic. Each cartridge is individually sealed in aluminum foil. Dispose of the cartridges after repeated use.
Catalog No. 50-189-9508 Supplier Electron Microscopy Sciences Supplier No. 7121202
These desiccant cartridges can be used in desiccator cabinets or non-vacuum desiccators, analytical balance cases, and precision laboratory instruments. The clear polystyrene dish allows for instant visual inspection of the silica gel color which changes from blue to pale pink as it absorbs moisture.
A highly porous membrane covers the dish of the desiccant providing good moisture absorption. The desiccant can be regenerated by heating cartridges in an oven at 50 degree C (122 degree F) overnight or in a 150 degree C (302 degree F) oven for one hour. The silica gel is inert and non-toxic. Each cartridge is individually sealed in aluminum foil. Dispose of the cartridges after repeated use.