Learn More
Hach Company Desiccant, Drierite (with indicator) 454g
Supplier:  Hach Company 2088701
Encompass
Indicating Drierite changes from a blue color when dry to red upon absorption of moisture. Packaged in 454-g (1 lb) containers.
Catalog No. 50-199-5299