Learn More
Chemglass Life Sciences Desiccant Flask Only, 24/40
SDP
Catalog No. CG129007
Encompass_Preferred
The desiccant flask has a 4mm glass stopcock and a serrated connection for the vacuum source. The flask and condenser joints are 24/40.
Catalog No. CG129007 Supplier Chemglass Life Sciences Supplier No. CG129007
The desiccant flask has a 4mm glass stopcock and a serrated connection for the vacuum source. The flask and condenser joints are 24/40.