Learn More
Synthware™ Desiccant Flask
GREENER_CHOICESDP
Catalog No. 31501060 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
34/45
Catalog No. Standard Taper Joint
31-501-060 34/45
1 options
Catalog No. 31-501-060 Supplier Synthware™ Supplier No. D133424
Add to Cart
Add to Cart
For use with D222434 drying chamber.
Specifications
Includes PTFE stopcock
For Use With (Application) Organic Synthesis
Type Desiccant Flask
Material Borosilicate Glass
For Use With (Equipment) Drying Chamber (D222434)
Quantity 1 Ea.
Standard Taper Joint 34/45