Learn More
Electron Microscopy Sciences Desiccant in A Cartridge 200 mm
SDP
Catalog No. 501899506
Encompass_Preferred
The desiccant in a cartridge regenerates repeatedly without degradation. These cartridges fit into the bottom of a standard glass or plastic desiccator. The cartridge consists of a durable long wearing polyester felt bag with a perforated Mylar cover sealed in place. The indicating gel beads will change their color from blue when dry to pink when moist. Saturated gel can be regenerated in a 150 degree C (300 degree F) vented oven for 3 hours or more (depending on size), until the blue color is restored.
Cartridges come individually sealed in a moisture-proof pouch ready to use.
Catalog No. 50-189-9506 Supplier Electron Microscopy Sciences Supplier No. 7120920
The desiccant in a cartridge regenerates repeatedly without degradation. These cartridges fit into the bottom of a standard glass or plastic desiccator. The cartridge consists of a durable long wearing polyester felt bag with a perforated Mylar cover sealed in place. The indicating gel beads will change their color from blue when dry to pink when moist. Saturated gel can be regenerated in a 150 degree C (300 degree F) vented oven for 3 hours or more (depending on size), until the blue color is restored.
Cartridges come individually sealed in a moisture-proof pouch ready to use.