Learn More
Perkin Elmer US LLC Desiccant Refills for PG-H2 Plus Generator
Catalog No. 509052366
Encompass_Preferred
These desiccant refills are for the PG-H2 Plus Pure Gas Hydrogen Generator and are sufficient for three refills. Pure gas hydrogen generators employ the newest membrane technology available for the safe production of pure hydrogen gas. This patented design is ideal for operation with gas analyzers, as fuel gas for flame tools, or as a source for pure hydrogen in plasma chambers and other isolated environments. Electrolytic membrane technology is preferred over alternative hydrogen generating techniques because it is clean, requires less maintenance and there is no need to store chemicals to maintain operation. The generators offer silent operation and require only deionized or distilled water with no caustic solutions which can affect the purity of the hydrogen.
Catalog No. 50-905-2366 Supplier Perkin Elmer US LLC Supplier No. N9306065
These desiccant refills are for the PG-H2 Plus Pure Gas Hydrogen Generator and are sufficient for three refills. Pure gas hydrogen generators employ the newest membrane technology available for the safe production of pure hydrogen gas. This patented design is ideal for operation with gas analyzers, as fuel gas for flame tools, or as a source for pure hydrogen in plasma chambers and other isolated environments. Electrolytic membrane technology is preferred over alternative hydrogen generating techniques because it is clean, requires less maintenance and there is no need to store chemicals to maintain operation. The generators offer silent operation and require only deionized or distilled water with no caustic solutions which can affect the purity of the hydrogen.