Learn More
Hach Company Desiccant Tube Filled with Molecular Sieve for AT/KF Titrator
Catalog No. NC3368690 Shop All Hach Company Products
Encompass
Desiccant tube filled with molecular sieve for use with AT/KF titrators.
  • Filled with molecular sieve.
  • Compatible with AT/KF titrators.
  • Listed as a one-time order; eligible for recurring orders.
  • Provides a replacement desiccant accessory for titration systems.
Catalog No. NC3368690 Supplier Hach Company Supplier No. LZE121
Desiccant tube filled with molecular sieve for use with AT/KF titrators.
  • Filled with molecular sieve.
  • Compatible with AT/KF titrators.
  • Listed as a one-time order; eligible for recurring orders.
  • Provides a replacement desiccant accessory for titration systems.