Learn More
Immunochemistry Technology Desiccant Packets 10/pk
Catalog No. NC1406351
Encompass
Desiccant Pack includes ten silica gel desiccant packets (5gm ea.) used to absorb moisture during immunoassay plate storage. Include a desiccant packet with stored ELISA plates to increase stability and shelf-life.
Catalog No. NC1406351 Supplier Immunochemistry Technology Supplier No. 6289
Desiccant Pack includes ten silica gel desiccant packets (5gm ea.) used to absorb moisture during immunoassay plate storage. Include a desiccant packet with stored ELISA plates to increase stability and shelf-life.