Learn More
Chemglass Life Sciences Dessiccator, 172mm I.D., 2.4L, Small Knob Top, 140mm Plate Dia, Complete
SDP
Catalog No. NC1682088
Encompass_Preferred
Dessiccator, 150mm I.D., 2.4L, Small Knob Top, 140mm Plate Dia, Complete. This 2.4L Pyrex Desiccator is designed for general analytical work. The sturdy knob makes cover easy to handle. Approximate bowl volume is 2,400mL. For replacement cover see CG-8266-R150CO. For lower part only, see CG-8266-R150LO. Please Note: Plate is not supplied and must be purchased seperately. WARNING: Do Not Heat Or Subject To Pressure. Not For Vacuum Applications.
Catalog No. NC1682088 Supplier Chemglass Life Sciences Supplier No. CG8266R150
Dessiccator, 150mm I.D., 2.4L, Small Knob Top, 140mm Plate Dia, Complete. This 2.4L Pyrex Desiccator is designed for general analytical work. The sturdy knob makes cover easy to handle. Approximate bowl volume is 2,400mL. For replacement cover see CG-8266-R150CO. For lower part only, see CG-8266-R150LO. Please Note: Plate is not supplied and must be purchased seperately. WARNING: Do Not Heat Or Subject To Pressure. Not For Vacuum Applications.