Learn More
Chemglass Life Sciences Desiccator, 223.5mm I.D., 5.8L, Large Top with 24/29 Stpk, 190mm Plate Dia, Complete
SDP
Catalog No. NC1483306
Encompass_Preferred
Desiccator, 200mm I.D., 5.8L, Large Top with 24/29 Stpk, 190mm Plate Dia, Complete. This 5.8L Pyrex desiccator has a rugged 24/29 standard taper stopcock with tooled hose connections that serve as the vacuum inlet. It is designed so that its manipulation is not affected by the vacuum. To introduce vacuum into the desiccator, loosen the stopcock to disengage the PTFE plug from contact with the glass. A half turn from this position closes the desiccator. This design eliminates the need for grease. The tubulation O.D. is approximately 10mm. Approximate bowl volume capacity is 5,800mL. For replacement bowl see CG-8266-R200LO. For replacement stopcock see CG-8272-RSTPK. Please Note: Plate is not supplied and must be purchased seperately. WARNING: Do Not Heat Or Subject To Pressure. Rated 1-Atmosphere Vacuum.
Catalog No. NC1483306 Supplier Chemglass Life Sciences Supplier No. CG8272R200
Desiccator, 200mm I.D., 5.8L, Large Top with 24/29 Stpk, 190mm Plate Dia, Complete. This 5.8L Pyrex desiccator has a rugged 24/29 standard taper stopcock with tooled hose connections that serve as the vacuum inlet. It is designed so that its manipulation is not affected by the vacuum. To introduce vacuum into the desiccator, loosen the stopcock to disengage the PTFE plug from contact with the glass. A half turn from this position closes the desiccator. This design eliminates the need for grease. The tubulation O.D. is approximately 10mm. Approximate bowl volume capacity is 5,800mL. For replacement bowl see CG-8266-R200LO. For replacement stopcock see CG-8272-RSTPK. Please Note: Plate is not supplied and must be purchased seperately. WARNING: Do Not Heat Or Subject To Pressure. Rated 1-Atmosphere Vacuum.