Learn More
Electron Microscopy Sciences Desiccator Cabinet 1
SDP
Catalog No. 50930900
Encompass_Preferred
Good for compact areas. It is see through from all sides. The door is air tight when closed and when it is opened it slides into a hide away compartment beneath the cabinet. Safely stackable up to five high. For compact areas, visual from all sides. Door is air tight and slides into a compartment under the cabinet. Stackable up to five. Inside: 8 1/4"(L) x 7"(W) x 5"(H) Outside: 9 1/4"(L) x 8-7/8"(W).
Catalog No. 50-930-900 Supplier Electron Microscopy Sciences Supplier No. 71215
Good for compact areas. It is see through from all sides. The door is air tight when closed and when it is opened it slides into a hide away compartment beneath the cabinet. Safely stackable up to five high. For compact areas, visual from all sides. Door is air tight and slides into a compartment under the cabinet. Stackable up to five. Inside: 8 1/4"(L) x 7"(W) x 5"(H) Outside: 9 1/4"(L) x 8-7/8"(W).