Learn More
Electron Microscopy Sciences Desiccator Cabinet 2
SDP
Catalog No. 50930901
Encompass_Preferred
Good for compact areas. It is see through from all sides. The door is air tight when closed and when it is opened it slides into a hide away compartment beneath the cabinet. Safely stackable up to five high. Comes with two shelves and vent holes, four sets of shelf rails for adjusting to low or high objects, "smoke" tinted in color, door opening is 9" wide and 8" high (23x20cm); inside depth is 9".
Catalog No. 50-930-901 Supplier Electron Microscopy Sciences Supplier No. 71216
Good for compact areas. It is see through from all sides. The door is air tight when closed and when it is opened it slides into a hide away compartment beneath the cabinet. Safely stackable up to five high. Comes with two shelves and vent holes, four sets of shelf rails for adjusting to low or high objects, "smoke" tinted in color, door opening is 9" wide and 8" high (23x20cm); inside depth is 9".