Learn More
Cole-Parmer Cole-Parmer Desiccator, PP/PC/neoprene, 10" dia
Catalog No. NC1467822 Shop All Cole-Parmer Products
Encompass_Preferred
Desiccator has a chemical-resistant polypropylene body and polycarbonate cover. A neoprene O-ring ffers a secure seal. Use grease on the flanges to make the unit completely airtight.
Catalog No. NC1467822 Supplier Cole-Parmer Supplier No. 0652526
Desiccator has a chemical-resistant polypropylene body and polycarbonate cover. A neoprene O-ring ffers a secure seal. Use grease on the flanges to make the unit completely airtight.