Learn More
Chemglass Life Sciences Desiccator, Polycarbonate, Size A
SDP
Catalog No. CG122610
Encompass_Preferred
Clear polycarbonate desiccator having extra heavy walls to eliminate the danger of implosion. Desiccator will hold a vacuum of 740mm of Hg (29”) for a period of 24 hours. Space saving design of the desiccator dome provides maximum interior clearances, giving an average of 13% greater interior volume than hemispherical domes. The supplied polypropylene stopcock has a PTFE plug which turns easily, allowing a slow, gentle entrance of air to prevent unnecessary turbulence and crucible damage. Stopcock port accepts 1/4” I.D. tubing. Supplied complete with a neoprene o-ring unaffected by temperatures down to 0ºC. Polypropylene plate that is supplied with the desiccator is 1/8” thick with 1/8” perforations for air transfer. The plate rests on the retaining ledge of the bottom section and has a center hole for easy removal.Plate Diameter: 140mmInside Diameter: 149mmClearance Above Plate: 121mmOverall Height: 206mmFlange O.D.: 171mm
Catalog No. CG122610 Supplier Chemglass Life Sciences Supplier No. CG122610
Clear polycarbonate desiccator having extra heavy walls to eliminate the danger of implosion. Desiccator will hold a vacuum of 740mm of Hg (29”) for a period of 24 hours. Space saving design of the desiccator dome provides maximum interior clearances, giving an average of 13% greater interior volume than hemispherical domes. The supplied polypropylene stopcock has a PTFE plug which turns easily, allowing a slow, gentle entrance of air to prevent unnecessary turbulence and crucible damage. Stopcock port accepts 1/4” I.D. tubing. Supplied complete with a neoprene o-ring unaffected by temperatures down to 0ºC. Polypropylene plate that is supplied with the desiccator is 1/8” thick with 1/8” perforations for air transfer. The plate rests on the retaining ledge of the bottom section and has a center hole for easy removal.Plate Diameter: 140mmInside Diameter: 149mmClearance Above Plate: 121mmOverall Height: 206mmFlange O.D.: 171mm