Learn More
Electron Microscopy Sciences Desiccator Sealing Rings, 100 mm
SDP
Catalog No. 501899549
Encompass_Preferred
Components for Dry Seal vacuum desiccators. These rings are molded from chemically resistant Viton rubber.
Size: 100 mm
Catalog No. 50-189-9549 Supplier Electron Microscopy Sciences Supplier No. 7126110
Components for Dry Seal vacuum desiccators. These rings are molded from chemically resistant Viton rubber.
Size: 100 mm