Learn More
Electron Microscopy Sciences Secador™ 1.0 Desiccator Cabinet, Clear
SDP
Catalog No. 501899514
Encompass_Preferred
This desiccator cabinet has a built-in hygrometer for quick check on RH levels. Transparent for easy viewing of contents. Superior chemical resistance and resists staining. Airtight with patented sealed construction. Stackable for a small footprint. Volume is 0.75cu ft, 1 shelf, size: 8.4"(H)x13.4"(W)x16.3"(D) (21.3x34.1x41.4cm)
Color: Clear
Catalog No. 50-189-9514 Supplier Electron Microscopy Sciences Supplier No. 7122500
This desiccator cabinet has a built-in hygrometer for quick check on RH levels. Transparent for easy viewing of contents. Superior chemical resistance and resists staining. Airtight with patented sealed construction. Stackable for a small footprint. Volume is 0.75cu ft, 1 shelf, size: 8.4"(H)x13.4"(W)x16.3"(D) (21.3x34.1x41.4cm)
Color: Clear