Learn More
Electron Microscopy Sciences Secador™ 4.0 Desiccator Horizontal, Blue
SDP
Catalog No. 501899530
Encompass_Preferred
Hygrometer, autodesiccator module with 2 latches. Volume of 2 cubic feet. Non-electric. Horizontal desicator with 3 shelves, 4 positions.
Color: Blue
Dimensions: 3.4"H x 20.4"W x 16.3"D
Catalog No. 50-189-9530 Supplier Electron Microscopy Sciences Supplier No. 7122807
Hygrometer, autodesiccator module with 2 latches. Volume of 2 cubic feet. Non-electric. Horizontal desicator with 3 shelves, 4 positions.
Color: Blue
Dimensions: 3.4"H x 20.4"W x 16.3"D