Learn More
Electron Microscopy Sciences Secador™ 4.0 Autodesiccator Horizontal, 115 V, Clear
SDP
Catalog No. 501899531
Encompass_Preferred
Hygrometer, autodesiccator module with 2 latches. Volume of 2 cubic feet. Reduces RH from 75% to <25% in 8 hours and maintains RH within narrow range. Automatic, continual desiccation. Two fans guarantee continuous airflow.
Color: Clear
Dimensions: 3.4"H x 20.4"W x 16.3"D
Catalog No. 50-189-9531 Supplier Electron Microscopy Sciences Supplier No. 7122815
Hygrometer, autodesiccator module with 2 latches. Volume of 2 cubic feet. Reduces RH from 75% to <25% in 8 hours and maintains RH within narrow range. Automatic, continual desiccation. Two fans guarantee continuous airflow.
Color: Clear
Dimensions: 3.4"H x 20.4"W x 16.3"D