Learn More
Electron Microscopy Sciences Secador™ 4.0 Autodesiccator Vertical, 115 V, Blue End Caps
SDP
Catalog No. 501899523
Encompass_Preferred
The superior dehumidification performance of the Autodesiccator model makes it a natural choice for more perishable and expensive products that require constant low humidity. Totally automatic, continual desiccation with silica beads, and generation every 20 minutes, keeps RH level at a constant level. No long regeneration lag between cycles-minimal change from opening the cabinet door. Two fans guarantee continuous airflow within the cabinet, and continuous exhaust of humid air.
Features a hygrometer and autodesiccator module. Has 3 shelves with 4 positions and 2 latches. Volume of 2 cubic feet. Reduces RH from 75% to <25% in 8 hours and maintains RH within narrow range. Automatic, continual desiccation. Two fans for continuous airflow.
Color: Blue End Caps
Catalog No. 50-189-9523 Supplier Electron Microscopy Sciences Supplier No. 7122716
The superior dehumidification performance of the Autodesiccator model makes it a natural choice for more perishable and expensive products that require constant low humidity. Totally automatic, continual desiccation with silica beads, and generation every 20 minutes, keeps RH level at a constant level. No long regeneration lag between cycles-minimal change from opening the cabinet door. Two fans guarantee continuous airflow within the cabinet, and continuous exhaust of humid air.
Features a hygrometer and autodesiccator module. Has 3 shelves with 4 positions and 2 latches. Volume of 2 cubic feet. Reduces RH from 75% to <25% in 8 hours and maintains RH within narrow range. Automatic, continual desiccation. Two fans for continuous airflow.
Color: Blue End Caps