Learn More
Hach Company Desiccator-Vacuum
Supplier:  Hach Company 2088800
Encompass
Round polycarbonate cover and polypropylene base create implosion-proof chamber at full vacuum Chamber is impervious to alkali and acidic conditions Recessed rim prevents grease (not included) from contacting bench surfaces when removed and eliminates need for a separate gasket This vacuum model features leak-proof stopcock Dimensions (height x diam) 245 mm x 330 mm (9 7in x 13in ) Holds 454 g (1 lb) of dessicant Ceramic dessicator plate (sold separately) is inert non-stick and corrosion resistant Quadrants crafted into plate for easy tracking of material locations Plate diameter 230 mm (9in )
Catalog No. 50-002-57123