Learn More
Bellco Glass, Inc. Desiccator Vacuum, Stopcock with PTFE spindle and Porcelain plate, 100 mm, Borosilicate
SDP
Supplier:  Bellco Glass, Inc. 452800741
Encompass_Preferred
Borosil Desiccator with cover and Porcelain Plate, Rubber Knob as per IS 6128 Borosilicate 3.3 glass, 300 mm, CS/1SpecificationsMaterial: 3.3 Borosilicate Glass Overall Dimension: 16.73" L x 16.73" W x 16.93" H3082045Class/Quality Grade: Type 1Steam Sterilization at 121º C is recommended for autoclavingNot recommended for heat applications
Catalog No. 50-255-0021