Learn More
Med Vet International Desiccators, 150 X 220MM, 2/Case
Catalog No. 503045704
Encompass_Preferred
Made of high-quality soda lime glass. The products enclose a porcelain plate which can hold the desiccant. The desiccant can absorb the water. Sizes refer to the inside diameter by total height.
Catalog No. 50-304-5704 Supplier Med Vet International Supplier No. GLQ150
Made of high-quality soda lime glass. The products enclose a porcelain plate which can hold the desiccant. The desiccant can absorb the water. Sizes refer to the inside diameter by total height.