Learn More
Med Vet International Desiccators, 210 X 320mm
Catalog No. 503046844
Encompass_Preferred
Made of high-quality soda lime glass. The products enclose a porcelain plate which can hold the desiccant. The desiccant can absorb the water. Sizes refer to the inside diameter by total height.
Catalog No. 50-304-6844 Supplier Med Vet International Supplier No. GLQ210
Made of high-quality soda lime glass. The products enclose a porcelain plate which can hold the desiccant. The desiccant can absorb the water. Sizes refer to the inside diameter by total height.