Learn More
Grainger QMark CWH3000 Series Wall Heater, 1,500W, 120V, 1 Phase, 1 ea
Catalog No. 502782673
Encompass_Preferred
This commercial-grade fan-forced wall heater provides efficient heating for versatile spaces including vestibules, lobbies, offices, and residential areas. Designed for recess or surface wall mounting, it features a durable steel grille and a permanently lubricated enclosed fan motor for reliable operation.
  • Automatic fan delay switch ensures airflow only after heating elements reach optimal temperature
  • Integral double-pole thermostat with a range of 40°F to 90°F
  • Manual reset thermal overheat protector for enhanced safety
  • Built-in power on/off switch
  • Includes fan-only operation mode for summer air circulation
  • Delivers 100 CFM of airflow with a capacity of 5,120 BTU/hr
Catalog No. 50-278-2673 Supplier Grainger Supplier No. 19PR51
This commercial-grade fan-forced wall heater provides efficient heating for versatile spaces including vestibules, lobbies, offices, and residential areas. Designed for recess or surface wall mounting, it features a durable steel grille and a permanently lubricated enclosed fan motor for reliable operation.
  • Automatic fan delay switch ensures airflow only after heating elements reach optimal temperature
  • Integral double-pole thermostat with a range of 40°F to 90°F
  • Manual reset thermal overheat protector for enhanced safety
  • Built-in power on/off switch
  • Includes fan-only operation mode for summer air circulation
  • Delivers 100 CFM of airflow with a capacity of 5,120 BTU/hr