Learn More
Cayman Chemical Desipramine-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 61361-34-0, 5 mg
SDP
Catalog No. 502909631
Encompass_Preferred
An internal standard for the quantification of desipramine by GC- or LC-MS
Catalog No. 50-290-9631 Supplier Cayman Chemical Supplier No. 368495 MG
An internal standard for the quantification of desipramine by GC- or LC-MS