Learn More
Medchemexpress LLC Desipramine-d4 | 61361-34-0 | 98.9% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB1272AS500UG
Encompass_Preferred
Desipramine-d4 is the deuterium labeled Desipramine. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to influence the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12948