Learn More
Medchemexpress LLC HY-B1272 100mg , Desipramine (hydrochloride) CAS:58-28-6 Purity:>98%
SDP
Catalog No. 502259404
Encompass_Preferred
HY-B1272 100mg Desipramine (hydrochloride) CAS: 58-28-6 Desipramine hydrochloride is an inhibitor of norepinephrine transporter (NET), 5-HT transporter (SERT) and dopamine transporter (DAT) with Kis of 4, 61 and 78,720 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9404 Supplier Medchemexpress LLC Supplier No. HYB1272100MG
HY-B1272 100mg Desipramine (hydrochloride) CAS: 58-28-6 Desipramine hydrochloride is an inhibitor of norepinephrine transporter (NET), 5-HT transporter (SERT) and dopamine transporter (DAT) with Kis of 4, 61 and 78,720 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.