Learn More
Medchemexpress LLC Desipramine hydrochloride | 58-28-6 | 99.8% | 500 MG
SDP
Catalog No. 5000412958
Encompass_Preferred
Desipramine hydrochloride is an inhibitor of norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT) with Ki values of 4, 61, and 78,720 nM, respectively. It is intended for research use only.
  • Inhibits norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT).
  • High purity of 99.76%.
  • Available in various quantities and formulations (solid, solution in DMSO).
  • Used in numerous publications, including studies on cell proliferation/viability, apoptosis-related proteins, and various cellular staining methods.
  • Relevant in research areas such as infection, neurological diseases, and cancer.
Catalog No. 50-004-12958 Supplier Medchemexpress LLC Supplier No. HYB1272500MG
Desipramine hydrochloride is an inhibitor of norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT) with Ki values of 4, 61, and 78,720 nM, respectively. It is intended for research use only.
  • Inhibits norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT).
  • High purity of 99.76%.
  • Available in various quantities and formulations (solid, solution in DMSO).
  • Used in numerous publications, including studies on cell proliferation/viability, apoptosis-related proteins, and various cellular staining methods.
  • Relevant in research areas such as infection, neurological diseases, and cancer.